Assay Details
| Gene Symbol: hbegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005161297.4 | XP_005161354.1 | 4 - 5 | 737 | 70 | |
| GenBank mRNA | BC066705.1 | - | 4 - 5 | 644 | 70 | |
| BC150223.1 | - | 4 - 5 | 712 | 70 | ||
More Information
Set Membership: |
|

hbegfb
Genomic Map