Assay Details
| Gene Symbol: hbegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100401.2 | NP_001093871.2 | 5 - 6 | 826 | 74 | |
| XM_005161297.4 | XP_005161354.1 | 5 - 6 | 825 | 74 | ||
| GenBank mRNA | BC066705.1 | - | 5 - 6 | 732 | 74 | |
| BC150223.1 | - | 5 - 6 | 800 | 74 | ||
| EH548388.1 | - | 3 - 4 | 372 | 74 | ||
| EH569504.1 | - | 1 - 2 | 848 | 74 | ||
More Information
Set Membership: |
|

hbegfb
Genomic Map