Assay Details
| Gene Symbol: wash1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110450.1 | NP_001103920.1 | 9 - 10 | 1311 | 108 | |
| GenBank mRNA | BC134941.1 | - | 9 - 10 | 1311 | 108 | |
More Information
Set Membership: |
|

wash1
Genomic Map