Assay Details
| Gene Symbol: stk24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110451.1 | NP_001103921.1 | 2 - 3 | 293 | 110 | |
| GenBank mRNA | BC134216.1 | - | 2 - 3 | 293 | 110 | |
More Information
Set Membership: |
|

stk24a
Genomic Map