Assay Details
| Gene Symbol: asmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110477.1 | NP_001103947.1 | 4 - 5 | 549 | 65 | |
| XM_009293101.3 | XP_009291376.1 | 4 - 5 | 606 | 65 | ||
| GenBank mRNA | BC133912.1 | - | 4 - 5 | 549 | 65 | |
More Information
Set Membership: |
|

asmt2
Genomic Map