Assay Details
| Gene Symbol: asmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110477.1 | NP_001103947.1 | 6 - 7 | 746 | 92 | |
| XM_009293101.3 | XP_009291376.1 | 6 - 7 | 803 | 92 | ||
| XM_009293103.3 | XP_009291378.1 | 2 - 3 | 379 | 92 | ||
| XM_021467210.1 | XP_021322885.1 | 2 - 3 | 313 | 92 | ||
| GenBank mRNA | BC133912.1 | - | 6 - 7 | 746 | 92 | |
More Information
Set Membership: |
|

asmt2
Genomic Map