Assay Details
| Gene Symbol: opn1lw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313715.1 | NP_001300644.1 | 1 - 2 | 210 | 77 | |
| GenBank mRNA | AF104904.1 | - | 1 - 2 | 209 | 77 | |
| AF109371.1 | - | 1 - 2 | 212 | 77 | ||
| BC059415.1 | - | 1 - 2 | 192 | 77 | ||
| EH435011.1 | - | 1 - 2 | 235 | 77 | ||
| EH444201.1 | - | 1 - 2 | 239 | 77 | ||
| KT008400.1 | - | 1 - 2 | 102 | 77 | ||
More Information
Set Membership: |
|

opn1lw1
Genomic Map