Assay Details
| Gene Symbol: pax3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131277.1 | NP_571352.1 | 7 - 8 | 1464 | 68 | |
| XM_017357647.2 | XP_017213136.2 | 7 - 8 | 1541 | 68 | ||
| XM_017357648.2 | XP_017213137.1 | 7 - 8 | 1465 | 68 | ||
| GenBank mRNA | AF014366.1 | - | 7 - 8 | 1464 | 68 | |
More Information
Set Membership: |
|

pax3a
Genomic Map