Assay Details
| Gene Symbol: acvr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131345.1 | NP_571420.1 | 9 - 10 | 1443 | 58 | |
| XM_021478751.1 | XP_021334426.1 | 8 - 9 | 1396 | 58 | ||
| GenBank mRNA | AF038425.1 | - | 9 - 10 | 1443 | 58 | |
| AF292028.1 | - | 7 - 8 | 1255 | 58 | ||
| BC162313.1 | - | 8 - 9 | 1308 | 58 | ||
| BC162317.1 | - | 8 - 9 | 1308 | 58 | ||
More Information
Set Membership: |
|

acvr1l
Genomic Map