Assay Details
| Gene Symbol: mapk12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131407.1 | NP_571482.1 | 2 - 3 | 361 | 68 | |
| GenBank mRNA | BC085415.1 | - | 2 - 3 | 361 | 68 | |
| BC165821.1 | - | 2 - 3 | 279 | 68 | ||
| DQ360071.1 | - | 2 - 3 | 245 | 68 | ||
| Y15075.1 | - | 329 | 68 | |||
More Information
Set Membership: |
|

mapk12a
Genomic Map