Assay Details
| Gene Symbol: mapk12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131407.1 | NP_571482.1 | 5 - 6 | 572 | 63 | |
| GenBank mRNA | BC085415.1 | - | 5 - 6 | 572 | 63 | |
| BC165821.1 | - | 5 - 6 | 490 | 63 | ||
| DQ360071.1 | - | 5 - 6 | 456 | 63 | ||
| Y15075.1 | - | 540 | 63 | |||
More Information
Set Membership: |
|

mapk12a
Genomic Map