Assay Details
| Gene Symbol: mapk12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131407.1 | NP_571482.1 | 8 - 9 | 804 | 76 | |
| GenBank mRNA | BC085415.1 | - | 8 - 9 | 804 | 76 | |
| BC165821.1 | - | 8 - 9 | 722 | 76 | ||
| DQ360071.1 | - | 8 - 9 | 688 | 76 | ||
| Y15075.1 | - | 772 | 76 | |||
More Information
Set Membership: |
|

mapk12a
Genomic Map