Assay Details
| Gene Symbol: actn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131522.2 | NP_571597.1 | 10 - 11 | 1258 | 73 | |
| GenBank mRNA | AY391405.1 | - | 10 - 11 | 73 | ||
| BC054911.1 | - | 10 - 11 | 1259 | 73 | ||
| FJ979801.1 | - | 10 - 11 | 73 | |||
More Information
Set Membership: |
|

actn3a
Genomic Map