Assay Details
| Gene Symbol: actn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131522.2 | NP_571597.1 | 13 - 14 | 1663 | 68 | |
| GenBank mRNA | BC054911.1 | - | 13 - 14 | 1664 | 68 | |
| FJ979801.1 | - | 13 - 14 | 1581 | 68 | ||
More Information
Set Membership: |
|

actn3a
Genomic Map