Assay Details
| Gene Symbol: actn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131522.2 | NP_571597.1 | 15 - 16 | 1990 | 77 | |
| GenBank mRNA | BC054911.1 | - | 15 - 16 | 1991 | 77 | |
More Information
Set Membership: |
|

actn3a
Genomic Map