Assay Details
| Gene Symbol: actn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131522.2 | NP_571597.1 | 18 - 19 | 2449 | 113 | |
| GenBank mRNA | AY391405.1 | - | 18 - 19 | 2426 | 113 | |
| BC054911.1 | - | 18 - 19 | 2450 | 113 | ||
| FJ979801.1 | - | 18 - 19 | 2367 | 113 | ||
More Information
Set Membership: |
|

actn3a
Genomic Map