Assay Details
| Gene Symbol: actn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131522.2 | NP_571597.1 | 20 - 21 | 2679 | 76 | |
| GenBank mRNA | BC054911.1 | - | 20 - 21 | 2680 | 76 | |
| FJ979801.1 | - | 20 - 21 | 2597 | 76 | ||
More Information
Set Membership: |
|

actn3a
Genomic Map