Assay Details
| Gene Symbol: actn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131522.2 | NP_571597.1 | 4 - 5 | 603 | 57 | |
| GenBank mRNA | AY391405.1 | - | 4 - 5 | 580 | 57 | |
| BC054911.1 | - | 4 - 5 | 604 | 57 | ||
| FJ979801.1 | - | 4 - 5 | 521 | 57 | ||
More Information
Set Membership: |
|

actn3a
Genomic Map