Assay Details
| Gene Symbol: actn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131522.2 | NP_571597.1 | 7 - 8 | 845 | 103 | |
| GenBank mRNA | AY391405.1 | - | 7 - 8 | 822 | 103 | |
| BC054911.1 | - | 7 - 8 | 846 | 103 | ||
| FJ979801.1 | - | 7 - 8 | 763 | 103 | ||
More Information
Set Membership: |
|

actn3a
Genomic Map