Assay Details
| Gene Symbol: actn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131522.2 | NP_571597.1 | 9 - 10 | 1028 | 89 | |
| GenBank mRNA | AY391405.1 | - | 9 - 10 | 1005 | 89 | |
| BC054911.1 | - | 9 - 10 | 1029 | 89 | ||
| FJ979801.1 | - | 9 - 10 | 946 | 89 | ||
More Information
Set Membership: |
|

actn3a
Genomic Map