Assay Details
| Gene Symbol: stx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131523.1 | NP_571598.1 | 7 - 8 | 539 | 68 | |
| GenBank mRNA | AF229154.1 | - | 7 - 8 | 539 | 68 | |
| BC076361.1 | - | 7 - 8 | 852 | 68 | ||
| BC165055.1 | - | 7 - 8 | 573 | 68 | ||
More Information
Set Membership: |
|

stx1b
Genomic Map