Assay Details
| Gene Symbol: bco2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131799.1 | NP_571874.1 | 4 - 5 | 569 | 117 | |
| GenBank mRNA | AJ290391.1 | - | 4 - 5 | 569 | 117 | |
| BC067696.1 | - | 2 - 3 | 195 | 117 | ||
| BC135024.1 | - | 4 - 5 | 582 | 117 | ||
More Information
Set Membership: |
|

bco2l
Genomic Map