Assay Details
| Gene Symbol: bco2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131799.1 | NP_571874.1 | 6 - 7 | 804 | 72 | |
| XM_021476337.1 | XP_021332012.1 | 6 - 7 | 848 | 72 | ||
| GenBank mRNA | AJ290391.1 | - | 6 - 7 | 804 | 72 | |
| BC067696.1 | - | 4 - 5 | 430 | 72 | ||
| BC135024.1 | - | 6 - 7 | 817 | 72 | ||
More Information
Set Membership: |
|

bco2l
Genomic Map