Assay Details
Gene Symbol: bco2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131799.1 | NP_571874.1 | 7 - 8 | 964 | 80 | |
XM_021476337.1 | XP_021332012.1 | 7 - 8 | 80 | |||
GenBank mRNA | AJ290391.1 | - | 7 - 8 | 964 | 80 | |
BC067696.1 | - | 5 - 6 | 590 | 80 | ||
BC135024.1 | - | 7 - 8 | 977 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
