Assay Details
| Gene Symbol: hnf1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131880.3 | NP_571955.3 | 1 - 2 | 503 | 81 | |
| XM_021481477.1 | XP_021337152.1 | 1 - 2 | 508 | 81 | ||
| GenBank mRNA | AF244140.2 | - | 1 - 2 | 446 | 81 | |
| AF430840.1 | - | 1 - 2 | 510 | 81 | ||
| AY336602.1 | - | 1 - 2 | 417 | 81 | ||
| AY569309.1 | - | 1 - 2 | 469 | 81 | ||
| BC129147.1 | - | 1 - 2 | 478 | 81 | ||
More Information
Set Membership: |
|

hnf1ba
Genomic Map