Assay Details
| Gene Symbol: hnf1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131880.3 | NP_571955.3 | 6 - 7 | 1490 | 74 | |
| GenBank mRNA | BC129147.1 | - | 6 - 7 | 1465 | 74 | |
More Information
Set Membership: |
|

hnf1ba
Genomic Map