Assay Details
| Gene Symbol: hnf1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131880.3 | NP_571955.3 | 7 - 8 | 1673 | 71 | |
| XM_021481477.1 | XP_021337152.1 | 6 - 7 | 1759 | 71 | ||
| XM_021481478.1 | XP_021337153.1 | 6 - 7 | 1463 | 71 | ||
| GenBank mRNA | AF244140.2 | - | 7 - 8 | 1616 | 71 | |
| AF430840.1 | - | 7 - 8 | 1680 | 71 | ||
| AY336602.1 | - | 7 - 8 | 1587 | 71 | ||
| AY569309.1 | - | 7 - 8 | 1640 | 71 | ||
| BC129147.1 | - | 7 - 8 | 1648 | 71 | ||
More Information
Set Membership: |
|

hnf1ba
Genomic Map