Assay Details
| Gene Symbol: acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153643.1 | NP_705929.1 | 10 - 11 | 1799 | 82 | |
| XM_009296804.3 | XP_009295079.1 | 10 - 11 | 1982 | 82 | ||
| XM_021469722.1 | XP_021325397.1 | 10 - 11 | 1473 | 82 | ||
| XM_021469723.1 | XP_021325398.1 | 10 - 11 | 1473 | 82 | ||
| XM_021469724.1 | XP_021325399.1 | 10 - 11 | 1457 | 82 | ||
| GenBank mRNA | AF435024.1 | - | 10 - 11 | 1799 | 82 | |
| AF435025.1 | - | 10 - 11 | 1411 | 82 | ||
| BC100043.1 | - | 10 - 11 | 1916 | 82 | ||
More Information
Set Membership: |
|

acvrl1
Genomic Map