Assay Details
| Gene Symbol: rps18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173234.1 | NP_775341.1 | 2 - 3 | 218 | 65 | |
| GenBank mRNA | AF210641.1 | - | 2 - 3 | 123 | 65 | |
| AY099517.1 | - | 2 - 3 | 218 | 65 | ||
| BC062289.1 | - | 2 - 3 | 199 | 65 | ||
More Information
Set Membership: |
|

rps18
Genomic Map