Assay Details
| Gene Symbol: eef1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173263.1 | NP_775370.1 | 3 - 4 | 247 | 63 | |
| GenBank mRNA | AY099512.1 | - | 3 - 4 | 247 | 63 | |
| AY394968.1 | - | 3 - 4 | 260 | 63 | ||
| BC058315.1 | - | 3 - 4 | 243 | 63 | ||
| BC066518.1 | - | 2 - 3 | 233 | 63 | ||
More Information
Set Membership: |
|

eef1g
Genomic Map