Assay Details
| Gene Symbol: rock2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174863.2 | NP_777288.1 | 17 - 18 | 2426 | 81 | |
| GenBank mRNA | AF295804.1 | - | 17 - 18 | 2426 | 81 | |
More Information
Set Membership: |
|

rock2a
Genomic Map