Assay Details
| Gene Symbol: rock2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174863.2 | NP_777288.1 | 19 - 20 | 2676 | 65 | |
| GenBank mRNA | AF295804.1 | - | 19 - 20 | 2676 | 65 | |
More Information
Set Membership: |
|

rock2a
Genomic Map