Assay Details
| Gene Symbol: rock2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174863.2 | NP_777288.1 | 26 - 27 | 3588 | 86 | |
| GenBank mRNA | AF295804.1 | - | 26 - 27 | 3588 | 86 | |
More Information
Set Membership: |
|

rock2a
Genomic Map