Assay Details
| Gene Symbol: mapk14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313759.1 | NP_001300688.1 | 2 - 3 | 481 | 63 | |
| XM_017358195.2 | XP_017213684.1 | 2 - 3 | 509 | 63 | ||
| GenBank mRNA | AB030898.1 | - | 2 - 3 | 491 | 63 | |
| BC063029.1 | - | 2 - 3 | 461 | 63 | ||
| BC063937.1 | - | 2 - 3 | 439 | 63 | ||
| EH447521.1 | - | 2 - 3 | 492 | 63 | ||
More Information
Set Membership: |
|

mapk14b
Genomic Map