Assay Details
| Gene Symbol: mapk14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313759.1 | NP_001300688.1 | 5 - 6 | 681 | 109 | |
| XM_017358195.2 | XP_017213684.1 | 5 - 6 | 709 | 109 | ||
| GenBank mRNA | AB030898.1 | - | 5 - 6 | 691 | 109 | |
| AY391436.1 | - | 5 - 6 | 609 | 109 | ||
| BC063029.1 | - | 5 - 6 | 661 | 109 | ||
| BC063937.1 | - | 5 - 6 | 639 | 109 | ||
| EH447521.1 | - | 5 - 6 | 692 | 109 | ||
More Information
Set Membership: |
|

mapk14b
Genomic Map