Assay Details
| Gene Symbol: mapk14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017358195.2 | XP_017213684.1 | 9 - 10 | 1031 | 106 | |
| GenBank mRNA | AB030898.1 | - | 9 - 10 | 1013 | 106 | |
More Information
Set Membership: |
|

mapk14b
Genomic Map