Assay Details
| Gene Symbol: birc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194397.3 | NP_919378.1 | 1 - 2 | 243 | 85 | |
| GenBank mRNA | AY057057.1 | - | 1 - 2 | 161 | 85 | |
| BC056739.1 | - | 1 - 2 | 236 | 85 | ||
| EH547926.1 | - | 1 - 2 | 268 | 85 | ||
| EH589060.1 | - | 1 - 2 | 233 | 85 | ||
More Information
Set Membership: |
|

birc5a
Genomic Map