Assay Details
| Gene Symbol: egfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194424.1 | NP_919405.1 | 16 - 17 | 2077 | 82 | |
| GenBank mRNA | AY154658.1 | - | 16 - 17 | 2077 | 82 | |
| AY332223.1 | - | 16 - 17 | 1982 | 82 | ||
More Information
Set Membership: |
|

egfra
Genomic Map