Assay Details
| Gene Symbol: egfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194424.1 | NP_919405.1 | 22 - 23 | 2860 | 73 | |
| GenBank mRNA | AY154658.1 | - | 22 - 23 | 2860 | 73 | |
| AY332223.1 | - | 22 - 23 | 2765 | 73 | ||
| BC163725.1 | - | 22 - 23 | 2731 | 73 | ||
More Information
Set Membership: |
|

egfra
Genomic Map