Assay Details
| Gene Symbol: egfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194424.1 | NP_919405.1 | 3 - 4 | 573 | 116 | |
| GenBank mRNA | AY154658.1 | - | 3 - 4 | 573 | 116 | |
| KT023304.1 | - | 3 - 4 | 409 | 116 | ||
More Information
Set Membership: |
|

egfra
Genomic Map