Assay Details
| Gene Symbol: idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199564.1 | NP_955858.1 | 191 | 61 | ||
| GenBank mRNA | BC048041.1 | - | 191 | 61 | ||
| BC063967.1 | - | 172 | 61 | |||
More Information
Set Membership: |
|

idh2
Genomic Map