Assay Details
| Gene Symbol: mfsd14a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199584.1 | NP_955878.1 | 10 - 11 | 1163 | 70 | |
| GenBank mRNA | BC056817.1 | - | 10 - 11 | 1163 | 70 | |
More Information
Set Membership: |
|

mfsd14a2
Genomic Map