Assay Details
| Gene Symbol: metap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199640.1 | NP_955934.1 | 7 - 8 | 888 | 64 | |
| GenBank mRNA | BC055563.1 | - | 7 - 8 | 888 | 64 | |
| BC068393.1 | - | 7 - 8 | 850 | 64 | ||
More Information
Set Membership: |
|

metap2b
Genomic Map