Assay Details
| Gene Symbol: gpd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199706.1 | NP_956000.1 | 1 - 2 | 96 | 73 | |
| GenBank mRNA | BC055382.1 | - | 1 - 2 | 96 | 73 | |
| BC067596.1 | - | 1 - 2 | 96 | 73 | ||
| DQ677583.1 | - | 1 - 2 | 86 | 73 | ||
More Information
Set Membership: |
|

gpd1b
Genomic Map