Assay Details
| Gene Symbol: wbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199710.1 | NP_956004.1 | 6 - 7 | 739 | 80 | |
| GenBank mRNA | BC053232.1 | - | 6 - 7 | 739 | 80 | |
More Information
Set Membership: |
|

wbp2nl
Genomic Map