Assay Details
| Gene Symbol: rraga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199714.1 | NP_956008.1 | 4 - 5 | 574 | 100 | |
| GenBank mRNA | BC049033.1 | - | 4 - 5 | 574 | 100 | |
More Information
Set Membership: |
|

rraga
Genomic Map