Assay Details
| Gene Symbol: hif1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310042.1 | NP_001296971.1 | 12 - 13 | 2358 | 72 | |
| GenBank mRNA | AY326951.1 | - | 12 - 13 | 2207 | 72 | |
| BC046875.1 | - | 11 - 12 | 1812 | 72 | ||
More Information
Set Membership: |
|

hif1ab
Genomic Map