Assay Details
| Gene Symbol: hif1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310042.1 | NP_001296971.1 | 4 - 5 | 848 | 70 | |
| GenBank mRNA | AY326951.1 | - | 4 - 5 | 697 | 70 | |
| BC046875.1 | - | 4 - 5 | 693 | 70 | ||
| BC165712.1 | - | 4 - 5 | 494 | 70 | ||
| EH457403.1 | - | 4 - 5 | 743 | 70 | ||
More Information
Set Membership: |
|

hif1ab
Genomic Map