Assay Details
| Gene Symbol: hif1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310042.1 | NP_001296971.1 | 8 - 9 | 1418 | 139 | |
| GenBank mRNA | AY326951.1 | - | 8 - 9 | 1267 | 139 | |
| BC046875.1 | - | 8 - 9 | 1263 | 139 | ||
| BC165712.1 | - | 8 - 9 | 1064 | 139 | ||
More Information
Set Membership: |
|

hif1ab
Genomic Map