Assay Details
| Gene Symbol: hif1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310042.1 | NP_001296971.1 | 9 - 10 | 1668 | 64 | |
| GenBank mRNA | AY326951.1 | - | 9 - 10 | 1517 | 64 | |
| BC046875.1 | - | 9 - 10 | 1513 | 64 | ||
| BC165712.1 | - | 9 - 10 | 1314 | 64 | ||
More Information
Set Membership: |
|

hif1ab
Genomic Map